Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ UV epi Light Module
GREENER_CHOICESDP

Catalog No. 01257188 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Wavelength:
254 nm
254 nm, 365 nm
302 nm
302 nm, 254 nm
302 nm, 365 nm
365 nm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength
01-257-188 302 nm, 365 nm
01-257-189 302 nm, 254 nm
01-257-191 254 nm, 365 nm
01-257-190 254 nm
01-257-187 302 nm
01-257-186 365 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-257-188 Supplier Syngene™ Supplier No. EPIMODMWLW
Add to Cart
Add to Cart

Controlled GeneSys software for all fluorescence requirements. Syngene™ UV epi Light module is an optional light module with high resolution camera. Gel documentation and analysis system

The new generation G:BOX with high resolution camera and application controlled GeneSys software for all fluorescence requirements.
Gel documentation and analysis system
  • Modular system
  • Light tight darkroom - suitable for fluorescence, upgradeable for chemiluminescence
  • 16bit 3.8MP camera with auto focus, auto exposure and series capture
  • Integration control - capture faint bands or features from any gel
  • Neutral fielding for background correction
  • Filter selection options for detection of a wide range of stains and dyes
  • GeneSys software includes a database of hundreds of applications
  • The system includes a motor driven zoom lens as standard and a motor driven filter wheel
  • Internal lighting comes from an optional UV transilluminator or a blue light LED model to further extend the applications possible with the G:BOX F3
  • With unlimited copies of GeneTools analysis software
Applications include:
  • Electrophoresis gels - Ethidium Bromide, Coomassie Blue, Silver stain, SYBR™ Gold, SYBR™ Green, SYBR™ Safe, GelStar™, SYPRO™ Red , SYPRO™ Ruby, SYPRO™ Orange, Fluorescein, Rhodamine Red™, Texas Red™, Pro-Q™ Diamond , Deep Purple™
  • Extended fluorescence - for use with EPI RGB lighting module
  • Agar plates
  • Cells in flasks
  • Autoradiographs
  • Membranes
  • Spots and slot blots
  • TLC plates
  • Microtitre plates
  • Macro-arrays

Specifications

Intensity Fixed
Wavelength 302 nm, 365 nm
Type UV epi Light, Mid Wave
For Use With (Equipment) G:BOX systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.