Fire Extinguishers
Filtered Search Results
Amerex™ 2.5 lb. Multipurpose Extinguisher
Contain specially fluidized and siliconized monoammonium-phosphate-based dry chemical
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Amerex™ 10 lb. (4.5kg) Multipurpose Extinguisher
Contains specially fluidized and siliconized monoammonium-phosphate-based dry chemical
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Includes | Wall mount bracket |
---|---|
UL Rating | 4A:80B:C |
Length (Metric) | 12.7 cm |
Capacity (English) | 10 lb. |
Product Type | Multi-purpose Fire Extinguisher |
Reagent Type | ABC dry chemical |
Capacity (Metric) | 4.5 kg |
Length (English) | 5 in. |
Temperature Range (English) | -65° to +120°F |
Material | Steel |
Temperature Range (Metric) | -53.8°C to +48.8°C |
Range | 15 to 21 ft. |
Discharge Time | 20 sec. |
Amerex™ Water Fire Extinguisher
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Amerex™ 5 lb. Multipurpose Extinguishers
Model B402
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Height (Metric) | 38.7 cm |
---|---|
Width (English) | 7.25 in. |
Length (Metric) | 10.8 cm |
Length (English) | 4.25 in. |
Material | Steel, Aluminum |
Range | 12 to 18 ft. |
Width (Metric) | 18.4 cm |
Dimensions (L x W x H) | 4.25 x 7.25 x 15.25 in. (10.8 x 18.4 x 38.7 cm) |
Product Type | Fire Extinguisher |
Reagent Type | Multipurpose ABC dry chemical |
Temperature Range (English) | -65° to +120°F |
Temperature Range (Metric) | -53.8°C to +48.8°C |
Discharge Time | 14 sec. |
Height (English) | 15.25 in. |
Amerex™ ABC Dry Chemical Fire Extinguishers
Rugged design with all metal valve construction
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Includes | Wall bracket |
---|---|
Product Type | Fire Extinguisher |
Reagent Type | ABC dry chemical |
Temperature Range (English) | -65° to +120°F |
Temperature Range (Metric) | -53.8°C to +48.8°C |
Range | 15 to 21 ft. |
Amerex™ Purple K Dry Chemical Fire Extinguishers
Fire-fighting agent for the oil, gas, chemical and utilities industries
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Product Type | Fire Extinguisher |
---|---|
Reagent Type | Purple K |
Temperature Range (English) | -65° to +120°F |
Temperature Range (Metric) | -53.8°C to +48.8°C |
Amerex™ Heavy-Duty Brackets for Fire Extinguishers
For use with a variety of Amerex fire extinguishers
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Material | Steel |
---|
Amerex™ CO2 Fire Extinguishers
Electrically nonconductive agent effective against Class B flammable liquid fires
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Includes | Wall mount bracket, hose and horn |
---|---|
Product Type | Fire Extinguisher |
Reagent Type | Carbon Dioxide |
Temperature Range (English) | -22° to +120°F |
Material | Aluminum |
Temperature Range (Metric) | -30°C to +48.8°C |
Range | 3 to 8 ft. |
Amerex™ Vehicle/Marine Bracket for 5 lb. Fire Extinguishers
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Essendant ProLine Pro 2.5 MP Fire Extinguisher, 1 A, 10 B:C, 100psi, 15h x 3.25 dia, 2.6lb
Be prepared; choose ProLine™ dry-chemical extinguishers for your commercial fire-suppression needs. Crystal-clear, step-by-step instruction label helps ensure effective use in a crisis. Easy-to-read, reliable pressure gauge simplifies inspections. Smooth-action, impact-resistant valve and lever ensure hassle-free operation. Lock-safe, easy-pull release pin minimizes accidental discharges without compromising emergency-reaction times. Rechargeable.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
MSC FIRE EXTINGUISHERS
Fire Extinguisher: Dry Chemical, 8" Dia, 30 lb Capacity
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
MSC FIRE EXTINGUISHER
Fire Extinguisher: 5" Dia
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
MSC 10LB CAP SURF MNT FIRE EXT CAB
NC3723049 10LB CAP SURF MNT FIRE EXT CAB
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
MLTPRPSE HME FIRE EXTINGUISHER
NC3233556 MLTPRPSE HME FIRE EXTINGUISHER
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More