Learn More
MEDPLUS SERVICES USA Super Rapid Readout Biological Indicator for Steam Sterilization
Supplier:  MEDPLUS SERVICES USA MMM 1492V
Encompass_Preferred
• Provides BI results in 1 hour • Offers 3X faster results than Attest Rapid BIs for 270°F and 275°F vacuum-assisted steam sterilization cycles • Helps deliver BI results in about the same time as it takes to cool a load • Meets ISO and FDA performance requirements for biological indicators
Catalog No. NC0796197
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MEDPLUS SERVICES USA Super Rapid Readout Biological Indicator for Steam Sterilization
Your feedback has been submitted: Thank you for helping us improve our website.