Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADI1 Antibody (OTI5C2), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271994PCP
Description
ADI1 Monoclonal specifically detects ADI1 in Human samples. It is validated for Flow Cytometry.Specifications
ADI1 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
ADI1 | |
Full length recombinant protein of human ADI1 (NP_060739) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI5C2 | |
Flow Cytometry | |
1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase, Acireductone dioxygenase (Ni(2+)-requiring), acireductone dioxygenase 1, APL1, ARDsubmergence induced protein 2, EC 1.13.11.53, FLJ10913, HMFT1638, membrane-type 1 matrix metalloproteinase cytoplasmic tail binding protein-1, Membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1, MTCBP1, MTCBP-1, Ni-ARD, SIPLMT1-MMP cytoplasmic tail-binding protein-1, Submergence-induced protein 2 homolog | |
Mouse | |
Affinity Purified | |
metabolism, Signal Transduction | |
55256.0 | |
Store at 4°C in the dark. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ADI1 Antibody (OTI5C2), PerCP, Novus Biologicals™ > 0.1 mL; PerCP
Spot an opportunity for improvement?Share a Content Correction