Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 Ε Rabbit anti-Bovine, Chicken, Human, Mouse, Rat, Rabbit, Sheep, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB9734
Description
This Anti-14-3-3 Ε Antibody, N-terminal is validated for use in western blot, immunohistochemistry for the detection of 14-3-3 Ε.
Specifications
14-3-3 ε N Terminal | |
Polyclonal | |
Liquid. | |
Rabbit | |
Serum | |
Primary | |
Bovine, Chicken, Human, Mouse, Rabbit, Rat, Sheep | |
Serum |
Immunohistochemistry, Western Blot | |
Unconjugated | |
YWHAE; KCIP-1; MDS; 14-3-3E; MDCR; FLJ45465 | |
Synthetic peptide from the N-terminal area of sheep 14-3-3 epsilon. | |
100 μL | |
NM_006761.3 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
14-3-3 Ε Rabbit anti-Bovine, Chicken, Human, Mouse, Rat, Rabbit, Sheep, Polyclonal, MilliporeSigma™ > 100μL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction