Learn More
aldolase B, fructose-bisphosphate, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ALDOB.
Supplier: Abnova Corporation H00000229A01
Description
Fructose-1,6-bisphosphate aldolase (EC 4.1.2.13) is a tetrameric glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Vertebrates have 3 aldolase isozymes which are distinguished by their electrophoretic and catalytic properties. Differences indicate that aldolases A, B, and C are distinct proteins, the products of a family of related 'housekeeping' genes exhibiting developmentally regulated expression of the different isozymes. The developing embryo produces aldolase A, which is produced in even greater amounts in adult muscle where it can be as much as 5% of total cellular protein. In adult liver, kidney and intestine, aldolase A expression is repressed and aldolase B is produced. In brain and other nervous tissue, aldolase A and C are expressed about equally. There is a high degree of homology between aldolase A and C. Defects in ALDOB cause hereditary fructose intolerance. [provided by RefSeq]
Sequence: DSQGKLFRNILKEKGIVVGIKLDQGGAPLAGTNKETTIQGLDGLSERCAQYKKDGVDFGKWRAVLRIADQCPSSLAIQENANASpecifications
aldolase B, fructose-bisphosphate | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant ALDOB. | |
ALDOB | |
ALDOB | |
ALDOB (NP_000026, 88 a.a. ∼ 170 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000035 | |
Mouse | |
50 μL | |
Yes | |
229 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.