Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANO7 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526428
Description
ANO7 may act as a calcium-activated chloride channel. This protein May play a role in cell-cell interactions.
Specifications
ANO7 | |
Polyclonal | |
ANO7 | |
D-TMPP, DTMPP, IPCA-5, IPCA5, NGEP, PCANAP5, PCANAP5L, TMEM16G, Dresden-transmembrane protein of the prostate, anoctamin-7, prostate cancer associated protein 5, transmembrane protein 16G | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50636 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q6IWH7 | |
ANO7 | |
KLH conjugated synthetic peptide between 106-135 amino acids from the N-terminal region of human ANO7 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ANO7 Rabbit anti-Human, Polyclonal, Invitrogen™ > 400 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction