Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CREB, Clone: LB9, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB5432/DEL

Description
ELISA, Western Blotting

Specifications
CREB | |
Unconjugated | |
Recombinant CREB-A protein | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 μg | |
Epigenetics | |
Human, Mouse, Rat | |
IgG1 |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MilliporeSigma™ anti-CREB, Clone: LB9, ™ (Chemicon™) > 100μg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction