Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EHMT1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 072046
Description
Use Anti-EHMT1 Antibody (Rabbit Polyclonal Antibody) validated in immunocytochemistry, ELISA, western blotting to detect EHMT1 also known as H3-K9-HMTase 5, Histone H3-K9 methyltransferase 5.
Specifications
EHMT1 | |
Polyclonal | |
Purified rabbit polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. | |
Rabbit | |
Protein A chromatography | |
Primary | |
Human | |
Purified |
ELISA, Immunocytochemistry, Western Blot | |
Unconjugated | |
DEL9q34; DKFZp667M072; EC 2.1.1.43; EUHMTASE1; Eu-HMTase1; FLJ12879; FP13812; GLP; GLP1; KIAA1876; KMT1D ; OTTHUMP00000022711; bA188C12.1 | |
The immunogen was a KLH-conjugated synthetic peptide corresponding to an internal region of human EHMT1. | |
100 μg | |
NP_079033 | |
2-8°C for 12 months Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial to pellet the solution. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction