Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPGD Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC248
Description
Detect Prostaglandin dehydrogenase 1 using this rabbit polyclonal antibody, Anti-HPGD Antibody validated for use in western blotting and immunohistochemistry.
Specifications
HPGD | |
Polyclonal | |
Purified rabbit Polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunohistochemistry, Western Blot | |
Unconjugated | |
HPGD; PGDH1 | |
KLH-conjugated linear peptide corresponding to human HPGD. | |
100 μg | |
NP_000851.2 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HPGD Rabbit anti-Human, Polyclonal, MilliporeSigma™ > 100μg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction