Learn More
ISG15 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA511269
Catalog No. PIPA511269
Description
Description
ISG15 is secreted from monocytes in response to type I interferons and causes natural killer (NK)-cell proliferation and an augmentation of non-MCH (major histocompatibility complex)-restricted cytotoxicity. Synthesis is stimulated by IFN-alpha or IFN-beta or IFN-omega , but not IFN-gamma . ISG15 expression is also induced by overexpression of interferon regulatory factors that participate in transcriptional regulation of IFN genes, and by influenza B virus. ISG15 is secreted also by cell lines of monocyte, T-lymphocyte, B-lymphocyte, human fibroblasts, and epithelial origins. The induction of terminal differentiation in human melanoma cells is associated with alterations in ISG15 expression. Enhancement of NK cell proliferation, augmentation of non-major histocompatibility complex-restricted cytotoxicity, and induction of IFN-gamma from T cells identify ISG15 as a member of the cytokine cascade and suggest that it may be responsible for amplifying and directing some of the immunomodulatory effects of IFN-alpha or IFN-beta. ISG15 has has also been shown to function intracellularly as a ubiquitin homolog.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISG15
Polyclonal
Unconjugated
P05161
ISG15
KLH conjugated synthetic peptide between 136-165 amino acids from the C-terminal region of human ISG15
400 μL
Primary
Human
Antibody
IgG
Western Blot
2.0 mg/mL
ISG15
Interferon-induced 17 kDa protein; Ubiquitin cross-reactive protein; hUCRP; Interferon-induced 15 kDa protein;G1P2, UCRP
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
9636
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ISG15 Rabbit anti-Human, Polyclonal, Invitrogen™ > 400 μL; Unconjugated
Your feedback has been submitted: Thank you for helping us improve our website.