Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF1 Antibody (1B3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197935H
Description
ARF1 Monoclonal specifically detects ARF1 in Human samples. It is validated for Western Blot, ELISA.Specifications
ARF1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ARF1 | |
Recombinant human ARF1 (1-181aa) purified from E. coli | |
100 μL | |
Primary | |
Anti-human ARF1 mAb, clone 1B3, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ARF1 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
1B3 | |
Western Blot, ELISA | |
ADP-ribosylation factor 1 | |
Mouse | |
Protein G purified | |
RUO | |
375 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ARF1 Antibody (1B3), HRP, Novus Biologicals™ > 100μL; HRP
Spot an opportunity for improvement?Share a Content Correction