Learn More
AnaSpec (ARG)9
Supplier:  AnaSpec AS61204
Encompass_Preferred
(Arg)9[RRRRRRRRR]; MW 1423.7; (1 mg) This is a peptide comprising of 9 arginine residues. It has been shown that poly-Arg peptides composed of 9 arginine residues are able to efficiently translocate across cells. It has also been shown in model systems that Arg-9 translocation involves nucleation of transient pores enabling flow of ions across the membrane. and that it's electrostatic attraction to the phosphate groups of membranes is a key property for its translocation.
Catalog No. 50-855-553
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
AnaSpec (ARG)9
Your feedback has been submitted: Thank you for helping us improve our website.