Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthcare Diagnostics Atellica IM TSH3-Ultra II
Click to view available options
Quantity:
130 Tests
650 Tests
Description
An assay for in vitro diagnostic use in the quantitative determination of thyroid-stimulating hormone (TSH, thyrotropin) in human serum and plasma.

Specifications
Specifications
Content And Storage | 5 ReadyPack primary reagent packs containing: • TSH3ULII Lite Reagent, solid phase • Ancillary Well Reagent • CAL3A H/L, 2 vials Store at 2°C to 8°C. |
CE Marker | Yes |
Clia Complexity | Moderate Complexity |
Detectable Analytes | Thyroid Stimulating Hormone (TSH) |
Detection Method | Chemiluminescence |
DoA Calibrators | Yes |
Sample Type | Serum, EDTA plasma, Heparinized plasma |
Type | Assay |
Disposable | Single-use |
Formulation | BSA, BGG, goat serum, sodium azide (<0.1%), surfactant, preservatives |
Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Siemens Healthcare Diagnostics Atellica IM TSH3-Ultra II >
Spot an opportunity for improvement?Share a Content Correction