Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG9A Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11056893PE
Description
ATG9A Polyclonal antibody specifically detects ATG9A in Human, Mouse, Rat, Alligator, Bovine, Chicken, Drosophila, Primate samples. It is validated for Flow CytometrySpecifications
ATG9A | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Alligator, Bovine, Chicken, Drosophila, Primate | |
Purified |
Flow Cytometry | |
PE | |
APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208 | |
A synthetic peptide made to a region within the C-terminus of human ATG9A (between residues 750-839). [Swiss-Prot: Q7Z3C6]. | |
0.1 mL | |
Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
79065 | |
Store at 4°C in the dark. | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ATG9A Antibody, PE, Novus Biologicals™ > 0.1 mL; PE
Spot an opportunity for improvement?Share a Content Correction