Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510

Specifications
Specifications
Product Type | BV480 Mouse Anti-Human CD11c |
Quantity | 25 Tests |
For Use With (Application) | Flow Cytometry |
Cross Reactivity | QC Testing: Human |
Form | BV480 |
Sample Volume | 5 μL |
Content And Storage | Store undiluted at 4°C |
Storage Buffer | Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD BV480 Mouse Anti-Human CD11c >
Spot an opportunity for improvement?Share a Content Correction