Learn More
Cancer Diagnostics, Inc.™ Vortex™ Biopsy Micro Cassettes
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Packaging Type:
Loose
Taped
Description
- Rounded micro hole interior basket without corners
- Filtration enhanced by side vents
- Two-piece lid design
- Processed biopsies are easy to find
- Various colors available
- Tubed, loose and taped options for packaging
Specifications
Specifications
Packaging Type | Loose |
Color | Pink |
Product Type | Biopsy Micro Cassette |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cancer Diagnostics, Inc.™ Vortex™ Biopsy Micro Cassettes >
Spot an opportunity for improvement?Share a Content Correction