Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHCHD4 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318483H
Description
CHCHD4 Polyclonal antibody specifically detects CHCHD4 in Human samples. It is validated for Western BlotSpecifications
CHCHD4 | |
Polyclonal | |
Western Blot | |
CHDH4, coiled-coil-helix-coiled-coil-helix domain containing 4, Coiled-coil-helix-coiled-coil-helix domain-containing protein 4, FLJ31709, MIA40mitochondrial intermembrane space import and assembly protein 40, mitochondrial intermembrane space import and assembly 40 homolog, TIMM40, translocase of inner mitochondrial membrane 40 homolog | |
Recognizes region between residues Between between 92 and 142.. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
131474 | |
Store at 4°C in the dark. | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CHCHD4 Antibody, HRP, Novus Biologicals™ > 0.1 mL; HRP
Spot an opportunity for improvement?Share a Content Correction