Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clostridium difficile Binary Toxin (B subunit), Chicken, HRP, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against Clostridium difficile Binary Toxin (B subunit).
Supplier: Abnova Corporation PAB29151
Specifications
Clostridium difficile Binary Toxin (B subunit) | |
Polyclonal | |
Chicken polyclonal antibody raised against Clostridium difficile Binary Toxin (B subunit). | |
In PBS, pH 7.2 (50% glycerol, 0.005% thimerosal). | |
Native purified Clostridium difficile Binary Toxin, B subunit with Freund's adjuvant. | |
100 μg | |
Primary | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
IgY |
ELISA | |
HRP | |
ELISA The optimal working dilution should be determined by the end user. | |
Chicken | |
Affinity purification | |
RUO | |
Clostridium species | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Clostridium difficile Binary Toxin (B subunit), Chicken, HRP, Polyclonal Antibody, Abnova™ > 100μg; HRP
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction