Learn More
United Scientific™ Cryo Coder For Skirted Base Vials Catalog No.: S15711
Only null left
Add to Cart
United Scientific™ Cryo Coder For Skirted Base Vials
SDPSCI_ED
Catalog No. p-200009350
Red
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Catalog No. S15711
Supplier: United Scientific™ P60104R
Only null left
Add to Cart
Only null left
Add to Cart
Easily identify different samples in your lab.
  • Designed to fit onto the top of our skirted base externally threaded cryogenic vial closures.
  • Provide quick visual identification of specific vials for ease in organizing inventory.
  • Available in a range of colors.
  • Compatible with United P60102 and P60103 skirted base cryo vials.
Specifications
Color Red
Type Cryo Coder
Vial Type For Use with United Skirted Base Cryo Vials
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
United Scientific™ Cryo Coder For Skirted Base Vials >
Your feedback has been submitted: Thank you for helping us improve our website.