Learn More
United Scientific™ Cryo Coder For Star and Round Base Vials
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Description
- Designed to fit onto the top of our star and round base cryogenic vial closures.
- Provide quick visual identification of specific vials for ease in organizing inventory.
- Available in a range of colors.
- Compatible with all United Scientific cryo vials except P60102 and P60103 skirted base cryo vials.
Specifications
Specifications
Color | White |
Type | Cryo Coder |
Vial Type | United Star and Round Base Cryo Vials |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
United Scientific™ Cryo Coder For Star and Round Base Vials >
Spot an opportunity for improvement?Share a Content Correction