Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Ligase IV Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5115128

Description
Antibody detects endogenous levels of total DNA Ligase IV.
The protein encoded by this gene is a DNA ligase that joins single-strand breaks in a double-stranded polydeoxynucleotide in an ATP-dependent reaction. This protein is essential for V(D)J recombination and DNA double-strand break (DSB) repair through nonhomologous end joining (NHEJ). This protein forms a complex with the X-ray repair cross complementing protein 4 (XRCC4), and further interacts with the DNA-dependent protein kinase (DNA-PK). Both XRCC4 and DNA-PK are known to be required for NHEJ. The crystal structure of the complex formed by this protein and XRCC4 has been resolved. Defects in this gene are the cause of LIG4 syndrome. Alternatively spliced transcript variants encoding the same protein have been observed.
Specifications
DNA Ligase IV | |
Polyclonal | |
Unconjugated | |
LIG4 | |
5830471N16Rik; DNA joinase; DNA ligase (ATP) 4; DNA ligase 4; DNA ligase IV; DNA repair enzyme; Lig4; LIG4S; ligase IV, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 4; polynucleotide ligase; sealase; tiny | |
Rabbit | |
Affinity chromatography | |
RUO | |
3981 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P49917 | |
LIG4 | |
A synthesized peptide derived from human LIG4(Accession P49917), corresponding to amino acid residues K626-P676. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNA Ligase IV Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction