Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Elastase, Porcine Pancreas, High Purity, Crystallized, Protein
Click to view available options
Quantity:
1000 U
250 U
Description
Native elastase from porcine pancreas. A serine protase that catalyzes the hydrolysis of proteins and peptides (especially at bonds adjacent to neutral amino acid residues), including albumin, casein, denatured collagen, elastin, fibrin, and hemoglobin and of a number of synthetic substrates containing aspartic acid, phenylalanine, or tyrosine. Inhibited by DFP, elastinal, and α2-macroglobulin.

Specifications
Specifications
Formulation | Lyophilized from 50mM Trehalose, 1mM Acetic Acid. |
Preparation Method | dH2O (10mg/mL) |
Quantity | 250 U |
Immunogen | Elastase |
Storage Requirements | Following reconstitution, elastase may be stored at 4°C at pH 6.0 for long term use. If incubated at room temperature at or near its optimum pH, elastase rapidly autolyzes to a mixture of peptides. Stock solutions are stable for up to 2 months at 4°C, pH 6.0. |
Regulatory Status | RUO |
Species | Porcine |
Recombinant | Native |
Form | Lyophilized |
Purity or Quality Grade | High Purity |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MilliporeSigma™ Elastase, Porcine Pancreas, High Purity, Crystallized, Protein >
Spot an opportunity for improvement?Share a Content Correction