Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calcitonin, Human, Synthetic, Peptide
Click to view available options
Quantity:
0.5 mg
Description
Synthetic, human calcitonin. A 32 amino acid peptide that is shown to stimulate bone formation and inhibit bone resorption.

Specifications
Specifications
Species | Human |
Purification Method | HPLC |
Purity | ≥97% |
Content And Storage | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 1 month at −20°C. |
Format | Lyophilized |
Formulation | Sold on the basis of peptide content. |
Product Type | Calcitonin |
Quantity | 0.5 mg |
Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MilliporeSigma™ Calcitonin, Human, Synthetic, Peptide >
Spot an opportunity for improvement?Share a Content Correction