Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514363
Description
This antibody is predicted to react with chicken, Xenopus and zebrafish based on sequence homology.
Homeobox-containing genes are thought to have a role in controlling development. In Drosophila, the 'engrailed' (en) gene plays an important role during development in segmentation, where it is required for the formation of posterior compartments. Different mutations in the mouse homologs, En1 and En2, produced different developmental defects that frequently are lethal. The human engrailed homologs 1 and 2 encode homeodomain-containing proteins and have been implicated in the control of pattern formation during development of the central nervous system.
Specifications
EN2 | |
Polyclonal | |
Unconjugated | |
EN2 | |
AUTS1; BB131122; EN2; En-2; engrailed 2; engrailed homeobox 2; engrailed homolog 2; engrailed-2; homeobox protein en-2; Homeobox protein engrailed-2; hu-En-2; mo-En-2; RGD1561842 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
13799, 2020 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P09066, P19622 | |
EN2 | |
KLH conjugated synthetic peptide between 243-271 amino acids from the C-terminal region of human EN2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EN2 Polyclonal Antibody, Invitrogen™ > 400 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction