Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
ENG Scientific Siderocyte Stain Kit, Potassium Ferrocyanide (2%) Catalog No.: ES3484
Only null left
Add to Cart

ENG Scientific Siderocyte Stain Kit, Potassium Ferrocyanide (2%)

Catalog No. p-3605942
2 x 250 mL
Click to view available options
Quantity:
1 x 250 mL
2 x 250 mL

Catalog No. ES3484

Supplier: ENG Scientific 3484

Only null left
Add to Cart
Only null left
Add to Cart

Used in Hematology. Siderocyte Stain Kit for Prussian Blue reaction. Siderotic granules stain bright blue.

  • Mixed with solution II to obtain Prussian blue reaction

Specifications

Quantity 2 x 250 mL
For Use With (Application) For in-vitro diagnostic use only
Packaging Bottles

For in vitro diagnostic use only. Due to product restrictions, please Sign In to purchase or view availability for this product.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ENG Scientific Siderocyte Stain Kit, Potassium Ferrocyanide (2%) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.