Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC1 Rabbit anti-Human, Clone: JM10-07, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP266823
Description
ERCC1 Monoclonal specifically detects ERCC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ERCC1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50-1:200 | |
COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
Rabbit | |
Protein A purified | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
2067 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
JM10-07 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
ERCC1 | |
Recombinant protein within Human ERCC1 aa 1-220/ 297. (SwissProt: P07992 Human; SwissProt: P07903 Mouse) | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ERCC1 Rabbit anti-Human, Clone: JM10-07, Novus Biologicals™ > 100μL
Spot an opportunity for improvement?Share a Content Correction