Learn More
International Enviroguard™ ViroGuard™ 2404 Disposable Coveralls with Hood and Boot Catalog No.: 17100883
Only undefined left
Add to Cart
International Enviroguard™ ViroGuard™ 2404 Disposable Coveralls with Hood and Boot
Catalog No. 17100883
4X-Large
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Catalog No. 17-100-883
Supplier: International Enviroguard™ 24044XL
Only undefined left
Add to Cart
Only undefined left
Add to Cart
ViroGuard™ Disposable Coveralls are designed and tested to protect against blood, blood-borne pathogens, bodily fluids, and many other liquid hazards. These coveralls boast elastic wrists and back, and a front zipper with storm flap.
  • Tear- and abrasion-resistant fabric protects against scrapes and scratches in the toughest of work environments
  • Fabric helps prevent heat stress with enhanced moisture vapor transmission, so you stay cool and comfortable on the job
  • With an attached hood and boots, you're protected from head-to-toe
  • Blue color makes stains and splashes less conspicuous
  • Possible applications include emergency response, category 3 black water, tank cleaning, crime scene remediation, and animal research
  • Fabric passes ASTM F1670 and F1671 tests for protection against blood, bodily fluids and blood borne pathogens
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure area
  • Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric
  • Elastic back stretches with user for more comfort and better tear resistance
Specifications
Product Type Protective Coveralls
Size 4X-Large
Gender Unisex
Material Microporous Laminate
Color Blue
Disposable Single-use
Waist Style Loose
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class Class 4
Resistance Bloodborne Pathogens, Liquid, Chemicals, Tears, Abrasion
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
International Enviroguard™ ViroGuard™ 2404 Disposable Coveralls with Hood and Boot >
Your feedback has been submitted: Thank you for helping us improve our website.