Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIV gp95 Antibody (SU1-30), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064378JF549
Description
FIV gp95 Monoclonal antibody specifically detects FIV gp95 in Virus samples. It is validated for Western Blot, Flow CytometrySpecifications
FIV gp95 | |
Monoclonal | |
Janelia Fluor 549 | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot, Flow Cytometry | |
SU1-30 | |
50mM Sodium Borate | |
Made to FIV gp95 | |
0.125 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FIV gp95 Antibody (SU1-30), Janelia Fluor™ 549, Novus Biologicals™ > 0.125 mL; Janelia Fluor 549
Spot an opportunity for improvement?Share a Content Correction