Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIF Antibody (016), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290227PE
Description
GIF Monoclonal antibody specifically detects GIF in Human samples. It is validated for ELISA, ELISASpecifications
GIF | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
2694 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
016 | |
ELISA, Sandwich ELISA Capture | |
gastric intrinsic factor (vitamin B synthesis), IFIntrinsic factor, IFMHgastric intrinsic factor, INF, TCN3 | |
Rabbit immunized with purified, recombinant Human GIF (Uniprot#: P27352-1, Met1-Tyr417) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GIF Antibody (016), PE, Novus Biologicals™ > 0.1 mL, PE
Spot an opportunity for improvement?Share a Content Correction