Learn More
Haws™ Eye/Face Wash Head
SDP
Haws™ Eye/Face Wash Head provides Zero Vertical Velocity™ stream engineering with even laminar flow for enhanced comfort, stability and effectiveness
$332.00 - $463.50
Specifications
Certifications/Compliance ANSI Z358.1
View More Specs
Products 3
Catalog Number Mfr. No. Flow Rate Material Price Quantity & Availability  
Catalog Number Mfr. No. Flow Rate Material Price Quantity & Availability  
18-888-254
SDP
View Documents
Haws™
SP65CTFP
3.7 gpm Plastic
Each for $339.00
Only null left
Add to Cart
18-888-255
SDP
View Documents
Haws™
SP65PD
4.2 gpm Plastic
Each for $332.00
Only null left
Add to Cart
18-888-256
SDP
View Documents
Haws™
SP65SS
3.7 gpm Stainless Steel
Each for $463.50
Only null left
Add to Cart
Description
Description
MSR eye/face wash head shall include inverted directional flow with laminar flow and an integral flow control, along with a yellow plastic pop-off dust cover.
Warranty and Services
1 year from ship date (Limited)
Specifications
Specifications
ANSI Z358.1
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haws™ Eye/Face Wash Head
Your feedback has been submitted: Thank you for helping us improve our website.