Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human COCO Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3047SP
This item is not returnable.
View return policy
Description
COCO/DAND5 Monoclonal specifically detects COCO/DAND5 in Human samples. It is validated for Western Blot.Greener Choice Claims
- Environmental benefits include:
- Non-Toxic
- Ozone-Safe or Safe for the Ozone Layer
- Renewable Energy
Specifications
COCO/DAND5 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q8N907 | |
DAND5 | |
E. coli-derived recombinant human COCO Arg23-Ala189 Accession # Q8N907 | |
25 μg | |
Primary | |
Detects human COCO in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) COCO, rmGremlin, rmPRDC, recombinant human (rh) DAN, rhCerberus, or recombinant chicken Caronte is observed. | |
Human | |
Purified |
Western Blot | |
322206 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
199699 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human COCO Antibody, R&D Systems™ > 25μg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction