Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Follistatin-like 1/FSTL1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB16941100
This item is not returnable.
View return policy
Description
Follistatin-like 1/FSTL1 Monoclonal specifically detects Follistatin-like 1/FSTL1 in Human samples. It is validated for ELISA.Specifications
Follistatin-like 1/FSTL1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Flik, FLJ50214, follistatin-like 1, Follistatin-like protein 1, Follistatin-related Protein, follistatin-related protein 1, FRP, FRPFLJ52277, FSL1, FSTL1, TSC-36 | |
Mouse myeloma cell line NS0-derived recombinant human Follistatin-like 1/FSTL1, Met1-Ile308, Accession No. Q12841 | |
100 μg | |
11167.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2b |
ELISA | |
229001R | |
ELISA | |
Q12841 | |
Rat | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Follistatin-like 1/FSTL1 in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human Follistatin-like 1/FSTL1 Antibody, R&D Systems™ > 100 μg; Unconjgated
Spot an opportunity for improvement?Share a Content Correction