Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ IL-10 Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair
$402.00 - $856.00
Specifications
Assay Range | 14.06 pg/mL |
---|---|
Assay Sensitivity | 14.06-900 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human IL-10.Specifications
14.06 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
IL10 | |
Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
IL-10 |
14.06-900 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes an antibody specific for Human IL-10. | |
Sandwich ELISA | |
3586 | |
Rabbit/Mouse | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 900 pg/mL is recommended. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ IL-10 Antibody Pair [HRP]