Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-15R alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $514.50
Specifications
Antigen | IL-15R alpha |
---|---|
Clone | 1020456 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | ||||||
MAB1472SP
![]() |
R&D Systems
MAB1472SP |
25 μg |
Each for $214.00
|
|
||||||
MAB1472100
![]() |
R&D Systems
MAB1472100 |
100 μg |
Each for $514.50
|
|
||||||
Description
IL-15R alpha Monoclonal specifically detects IL-15R alpha in Human samples. It is validated for Immunohistochemistry.Specifications
IL-15R alpha | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD215, CD215 antigen, IL15 R alpha, IL-15 R alpha, IL-15 receptor subunit alpha, IL15RA, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
Human embryonic kidney cell HEK293-derived human IL-15R alpha, Met1-Thr205, Accession # Q13261 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1020456 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q13261 | |
3601 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human IL-15R alpha Antibody, R&D Systems™