Learn More
BioMed Diagnostics Dermatophyte fungi growth and detection of distinct colony morphology and color. | Detects: Trichophyton mentagrophytes, T. rubrum, T. tonsurans, and Microsporum gypseum. | Available in: 20 pack
Supplier: BioMed Diagnostics 11263002
InTray SAB-FungID is a dynamic system with built-in components and features that are designed for user compatibility and ease of dermatophyte fungi detection (i.e., 10x Obj. microscopy direct from the tray). The medium allows for the growth & observation of distinct colony morphology and color (i.e., pigments) of dermatophyte fungi, while inhibiting most gram-positive bacteria, gram-negative bacteria, yeast and saprophytic fungi. | Detects: Trichophyton mentagrophytes, T. rubrum, T. tonsurans, and Microsporum gypseum. | Time to Result: 1 -14 days, incubated at 25C for positive for Dermatophyte identification. | Available in: 20 pack
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.