Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL5/CD158f Antibody (UP-R1), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262222MFV500
Description
KIR2DL5/CD158f Monoclonal antibody specifically detects KIR2DL5/CD158f in Human samples. It is validated for Flow CytometrySpecifications
KIR2DL5/CD158f | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
UP-R1 | |
Flow Cytometry | |
CD158F, CD158F1, killer cell immunoglobulin-like receptor 2DL5A, killer cell immunoglobulin-like receptor KIR2DL5A, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A, killer-cell Ig-like receptor, KIR2DL5, KIR2DL5.1, KIR2DL5.3, KIR2DL5B | |
Human CD158f-Ig fusion protein | |
0.1 mL | |
Immunology | |
57292 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KIR2DL5/CD158f Antibody (UP-R1), mFluor Violet 500 SE, Novus Biologicals™ > 0.1 mL; mFluor Violet 500 SE
Spot an opportunity for improvement?Share a Content Correction