Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KvBeta2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310368100UL
Description
KvBeta2 Polyclonal specifically detects KvBeta2 in Human samples. It is validated for Western Blot.Specifications
KvBeta2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
HKvbeta2, HKvbeta2.1, HKvbeta2.2, K(+) channel subunit beta-2, K+ channel beta-2 subunit, KCNA2BAKR6A5, KCNK2, KV-BETA-2, MGC117289, potassium channel shaker chain beta 2, potassium voltage-gated channel, shaker-related subfamily, beta member 2, voltage-gated potassium channel subunit beta-2 | |
The immunogen is a synthetic peptide directed towards the middle region of Human KvBeta2. Peptide sequence KIFWGGKAETERGLSRKHIIEGLKASLERLQLEYVDVVFANRPDPNTPME | |
100 μg | |
Signal Transduction | |
8514 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KvBeta2 Rabbit anti-Human, Polyclonal, Novus Biologicals™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction