Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRG1 Antibody (LRG1/4883), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314263B
This item is not returnable.
View return policy
Description
LRG1 Monoclonal antibody specifically detects LRG1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
LRG1 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2c κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
LRG1/4883 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
1300008B03Rik, 2310031E04Rik, HMFT1766, leucine-rich alpha-2-glycoprotein, leucine-rich alpha-2-glycoprotein 1, LRGleucine rich alpha 2 glycoprotein | |
Recombinant fragment (around aa 30-180) of human LRG1 protein (exact sequence is proprietary) (Uniprot: P02750) | |
0.1 mL | |
Cancer | |
116844 | |
Store at 4°C in the dark. | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LRG1 Antibody (LRG1/4883), Biotin, Novus Biologicals™ > 0.1 mL; Biotin
Spot an opportunity for improvement?Share a Content Correction