Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TROY Monoclonal Antibody (19), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA529834
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human TNFRSF19/TROY.
TROY can mediate activation of c-Jun and NF-kappa-B. May promote caspase-independent cell death. Isoform 2 and isoform 3 may act as decoy receptors.
Specifications
TROY | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9NS68 | |
Tnfrsf19 | |
Recombinant Human TNFRSF19/TROY Protein (Met1-Leu170) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
19 | |
Unconjugated | |
Tnfrsf19 | |
AL023044; AW123854; RGD1564996; TAJ; TAJ-alpha; TNF receptor superfamily member 19; Tnfrsf19; Toxicity and JNK inducer; TRADE; TROY; tumor necrosis factor receptor superfamily member 19; tumor necrosis factor receptor superfamily, member 19; UNQ1888/PRO4333 | |
Mouse | |
Protein A | |
RUO | |
55504 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TROY Monoclonal Antibody (19), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction