Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Methylene Blue Solution II, 0.05 mL (1 drop) = 0.1 mg/L Sulfide, Ricca Chemical Catalog No.: 49104
Only undefined left
Add to Cart

Methylene Blue Solution II, 0.05 mL (1 drop) = 0.1 mg/L Sulfide, Ricca Chemical
SDP

Catalog No. p-3251948
120 mL, Natural Poly Bottle
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle

Catalog No. 4910-4

Supplier: Ricca Chemical Company 49104

Only undefined left
Add to Cart
Only undefined left
Add to Cart

Specifications

Chemical Name or Material Methylene Blue Solution II
CAS 7220-79-3, 7732-18-5
CAS Min % 98.0, 0.01
CAS Max % 100.0, 0.01
Grade Laboratory
Packaging Natural Poly Bottle
Color Blue
Quantity 120 mL
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methylene Blue Solution II, 0.05 mL (1 drop) = 0.1 mg/L Sulfide, Ricca Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.