Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Watson Gloves Moscow Mule Sherpa Lined Gloves Catalog No.: 17099770
Only null left
Add to Cart

Watson Gloves Moscow Mule Sherpa Lined Gloves

Catalog No. 17099770
Large
Click to view available options
Size:
Large
X-Large

Catalog No. 17-099-770

Supplier: Watson Gloves 9200IL

Only null left
Add to Cart
Only null left
Add to Cart

Moscow Mule Sherpa Lined Gloves have heavy sherpa lining, Dryhide™ water resistant full-grain cowhide leather palm and pigskin leather back, shirred elastic wrist with Velcro™ closure and extended gauntlet style cuff.

  • Heavy sherpa lining keeps hands warm in cold conditions
  • Dryhide™ water resistant full-grain cowhide leather palm
  • Breathable pigskin leather back
  • ’’Be safe, be seen’’ with 2 in. reflective strip on cuff
  • Extended gauntlet style cuff
  • Shirred elastic wrist
  • Velcro™ closure

Specifications

Construction Cut and Sewn
Description Sherpa-lined Glove
Coating Material Uncoated
Color Black
Exterior Finish Smooth
Hand Ambidextrous
Material Pigskin Leather
Product Type Cold-resistant Gloves
Size Large
For Use With (Application) General Winter, Industrial, Refrigeration, General Labor
Liner Material Heavy Sherpa
Lining Lined
Series Moscow Mule
Length (English) 13.5 in.
Length (Metric) 34 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Watson Gloves Moscow Mule Sherpa Lined Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.