Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Avidin from Egg White
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg
Description
Avidin was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding.
- CAS Number: 1405-69-2
- EC Number: 215-783-6
- MDL Number: MFCD00130572
- Format: White Crystalline Powder
- Specific Activity: Approx. 10 to 15 units/mg protein
- Soluble in water or dilute aqueous buffer
- Isoelectric point: 10

Specifications
Specifications
Quantity | 10 mg |
Source | Chicken |
Content And Storage | 0°C |
Form | Lyophilized |
For Use With (Application) | Molecular Biology |
Product Type | Avidin from Egg White |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Avidin from Egg White >
Spot an opportunity for improvement?Share a Content Correction