Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
MP Biomedicals™ Avidin from Egg White Catalog No.: ICN15040701
Only null left
Add to Cart

MP Biomedicals™ Avidin from Egg White

Catalog No. p-4605317
1 mg
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg

Catalog No. ICN15040701

Supplier: MP Biomedicals™ 0215040701

Only null left
Add to Cart
Only null left
Add to Cart

Avidin is a basic glycoprotein consisting of four essentially identical subunits

Avidin was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding.

  • CAS Number: 1405-69-2
  • EC Number: 215-783-6
  • MDL Number: MFCD00130572
  • Format: White Crystalline Powder
  • Specific Activity: Approx. 10 to 15 units/mg protein
  • Soluble in water or dilute aqueous buffer
  • Isoelectric point: 10

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Source Chicken
Content And Storage 0°C
Form Lyophilized
For Use With (Application) Molecular Biology
Product Type Avidin from Egg White
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Avidin from Egg White >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.