Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL17 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318407C
Description
MRPL17 Polyclonal antibody specifically detects MRPL17 in Human samples. It is validated for Western BlotSpecifications
MRPL17 | |
Polyclonal | |
Western Blot | |
L17mt, LIP2, LYST-interacting protein 2, LYST-interacting protein LIP2, mitochondrial ribosomal protein L17, MRP-L17, MRP-L26, RPL17L, RPML26,39S ribosomal protein L17, mitochondrial | |
Recognizes region between residues Between Between 25 and 75.. | |
0.1 mL | |
Stem Cell Markers | |
63875 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MRPL17 Antibody, DyLight 650, Novus Biologicals™ > 0.1 mL; DyLight 650
Spot an opportunity for improvement?Share a Content Correction