Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Invitrogen™ Novex™ Sharp Pre-stained Protein Standard Catalog No.: LC5800
Only null left
Add to Cart

Invitrogen™ Novex™ Sharp Pre-stained Protein Standard

Catalog No. lc5800
2 x 250 μL
Encompass_Preferred
Click to view available options
Quantity:
2 x 250 μL

Catalog No. LC5800

Supplier: Invitrogen™ LC5800

Only null left
Add to Cart
Only null left
Add to Cart

Includes

Novex Sharp Pre-stained Protein Standards provided as 2 x 250μL (total of 50 applications of 10μL each) of ready-to-use standardmixture Novex Sharp Pre-Stained standard loading buffer consists of 65mM Tris pH 6.5, 30% glycerol, 2% SDS, and 2.5mM ETDA

Designed for accurate, easy, and convenient molecular weight estimation of a wide range of molecular weight proteins during SDS-PAGE and Western blotting

  • Accurate - sharp bands for the accurate estimation of molecular weight
  • Unambiguous - each band in the standard is pre-stained with a unique color for easy interpretation of results
  • Comprehensive - a wide range of molecular weight bands consisting of 12 proteins in the range of 3.5 to 260kDa
  • Convenient - a ready-to-use formulation eliminates the need to heat, reduce, or add sample buffer prior to use
  • Adaptable - suitable for most gel types, recommended for use with Novex™ NuPAGE™, Tris-Glycine, and Tricine gels
  • Gel Compatibility: Bolt Bis-Tris Plus Gels, NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels

The Novex Sharp Protein Standard is also available in an unstained format.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Novex™ Sharp Pre-stained Protein Standards are provided as 2 x 250 μL (total of 50 applications of 10 μL each) of ready-to-use standard mixture. The Novex™ Sharp Pre-Stained standard loading buffer consists of 65 mM Tris pH 6.5, 30% glycerol, 2% SDS, and 2.5 mM ETDA.

Store at -20°C.
Detection Method Colorimetric
Number of Markers 12
Product Type Protein Ladder
Ready to Load Yes
Size Range 3.5 to 260 kDa
Stain Type 3 colors: Pink, Yellow, Blue
Gel Compatibility Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels
Molecular Weight (g/mol) 260, 160, 110, 80, 60, 50, 40, 30, 20, 15, 10, 3.5 kDa
Product Line Novex™
Quantity 2 x 250 μL
Shipping Condition Approved for shipment on Wet or Dry Ice
System Type Western Blotting, SDS-PAGE
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Invitrogen™ Novex™ Sharp Pre-stained Protein Standard

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.