Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human Centrin 1 His Protein

Click to view available options
:
50μg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-172 of Human Centrin 1 The Recombinant Human Centrin 1 Protein is derived from E. coli. The Recombinant Human Centrin 1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | Q12798 |
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 0.1M NaCl, 10% glycerol,1mM DTT |
Gene ID (Entrez) | 1068 |
Molecular Weight (g/mol) | 22.1kDa |
Name | Centrin 1 Protein |
Purification Method | SDS-PAGE |
Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
Cross Reactivity | Human |
Purity or Quality Grade | >90% |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human Centrin 1 His Protein >
Spot an opportunity for improvement?Share a Content Correction