Learn More
Dunlop™ Onguard™ Goliath™ PVC Knee Boots
Ideal for general industrial and safety applications
Supplier:  Dunlop™ 89682 13
 View more versions of this product
Catalog No. 19-099-167J
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Constructed of a special urethane/PVC compound with Elvaloy™ and polymeric plasticizers for enhanced strength and high abrasion resistance
  • Superior protection from animal fats (especially poultry), acids, oils, greases, and other chemicals
  • Net-lined for easy on and off, with a removable insole
  • Built-in steel shank for firm arch support
  • Black with a gray Power-Lug outsole
Order Info
Available with or without steel toe in men's sizes only
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASTM F2413-05
Pull-on
16 in.
Steel
Gray power-lug outsole and removable insole
Safety Steel Toe
Yes
Black
Used in food processing, chemical plants, lawn care, canneries, poultry processing, fish processing, breweries and mining
Urethane, PVC
13
Steel
Animal fats, acids, oils, greases and chemicals
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dunlop™ Onguard™ Goliath™ PVC Knee Boots > Steel Safety toe; Men's; Size: 13
Your feedback has been submitted: Thank you for helping us improve our website.