Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577667
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Receptor for UTP and UDP coupled to G-proteins that activate a phosphatidylinositol-calcium second messenger system. Not activated by ATP or ADP.
Specifications
P2Y4 | |
Polyclonal | |
Unconjugated | |
P2RY4 | |
P2YR4, P2P | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5030, 57385, 63843 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.9 mg/mL | |
PBS with 1% BSA, 5% sucrose and 0.05% sodium azide; pH 7.4 | |
O35811, P51582, Q9JJS7 | |
P2ry4 | |
Peptide (C)HEES ISRWA DTHQD, corresponding to residues 337-350 of rat P2Y4 | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
P2Y4 Polyclonal Antibody, Invitrogen™ > 50 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction