Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phafin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5116236

Description
APPD (Apoptosis-inducing protein D; phafin 1) is predicted through gene annotation and contains pleckstrin homology domain. It may binds to two Zn++ ions through its FYVE zinc finger domain. Biological function of this protein has not been clearly mapped yet.
Specifications
Phafin 1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96S99 | |
PLEKHF1 | |
KLH conjugated synthetic peptide between 51-79 amino acids from the N-terminal region of human PLEKHF1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PLEKHF1 | |
1810013P09Rik; AI037048; Apoptosis-inducing protein; apoptosis-inducing protein D; APPD; Lapf; lysosome-associated apoptosis-inducing protein containing PH and FYVE domains; PH and FYVE domain-containing protein 1; PH domain-containing family F member 1; phafin 1; PHAFIN1; phafin-1; pleckstrin homology and FYVE domain containing 1; pleckstrin homology domain containing, family F (with FYVE domain) member 1; pleckstrin homology domain-containing family F member 1; Plekhf1; SGP-1; ZFYVE15; Zinc finger FYVE domain-containing protein 15 | |
Rabbit | |
Protein A | |
RUO | |
79156 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phafin 1 Polyclonal Antibody, Invitrogen™ > 400 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction